missing translation for 'onlineSavingsMsg'
Learn More

BD Cy™5 Annexin V

Product Code. p-4376435
Click to view available options
Quantity:
100 Tests
200 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15859828

Brand: BD 559933

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sensitive probe for identifying apoptotic cells, binding to negatively charged phospholipid surfaces (Kd of ∽5 x 102) with a higher affinity for phosphatidylserine (PS) than most other phospholipids

Apoptosis is a normal physiologic process which occurs during embryonic development as well as in maintenence of tissue homeostasis. The apoptotic program is characterized by certain morphologic features, including loss of plasma membrane asymmetry and attachment, condensation of cytoplasm and nucleus, and internucleosomal cleavage of DNA. Loss of plasma membrane is one feature. In apoptotic cells, membrane phospholipid phosphatidylserine (PS) is translocated from inner to outer leaflet of plasma membrane, thereby exposing PS to external cellular environment. Annexin V is a 35-36 kDa Ca2+ dependent phospholipid-binding protein that has a high affinity for PS, and binds to cells with exposed PS. Annexin V may be conjugated to fluorochromes including Cy5. This format retains its high affinity for PS and thus serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis. Since externalization of PS occurs in earlier stages of apoptosis, Cy5 Annexin V staining can identify apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation. Cy5 Annexin V staining precedes loss of membrane integrity which accompanies latest stages of cell death resulting from either apoptotic or necrotic processes. Therefore, staining with Cy5 Annexin V is typically used in conjunction with a vital dye such as propidium iodide (PI) or 7-Amino-Actinomycin (7-AAD) to allow investigator to identify early apoptotic cells (7-AAD negative, Cy5 Annexin V positive)

  • Format: Cy5
  • Cy5 Annexin V staining precedes the loss of membrane integrity
  • Used in conjunction with a vital dye such as propidium iodide (PI) or 7-Amino-Actinomycin (7-AAD)
  • Emission wavelength: 670nm
  • Excitation wavelength: 625 - 650nm

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow Cytometry
Formulation Stored in Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Name Cy™5 Annexin V
Quantity 100 Tests
Storage Requirements 4°C
Conjugate Cy5
Form Liquid

Research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.