missing translation for 'onlineSavingsMsg'
Learn More

BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution Kit

Product Code. 10482735
Change view
Click to view available options
Quantity:
250 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10482735 250 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10482735 Supplier BD Supplier No. 554714
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Fixation/Permeabilization solution (125mL)
  • Perm/Wash Buffer, 10x concentrate containing Fetal Bovine Serum (FBS) and saponin (100mL)

Enables the fixation and permeabilization of cells which is necessary for staining intracellular cytokines with fluorochrome-conjugated anti-cytokine antibodies

  • Provides two reagents, fixation/permeabilization solution and Perm/Wash Buffer
  • After cell fixation and permeabilization, the Perm/Wash Buffer is used to wash the cells and to dilute the anti-cytokine antibodies for staining

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Includes 125 mL fixation/permeabilization solution, perm/Wash™ buffer, fetal bovine serum (FBS) 10X and 100 mL saponin
No. of Tests 250
Quantity 250 Tests
Regulatory Status RUO
Product Type Fixation/Permeabilization Reagent
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BD Cytofix/Cytoperm Fixation/Permeabilization Solution Kit
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Formaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.