missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Functional Beads
Click to view available options
Product Type:
Functional Bead (A4)
Functional Bead (A5)
Functional Bead (A6)
Functional Bead (A9)
Functional Bead (B5)
Functional Bead (B7)
Functional Bead (B8)
Functional Bead (C4)
Functional Bead (C6)
Functional Bead (D4)
Functional Bead (D8)
Functional Bead (E4)
Functional Bead (E8)
Unit Size:
Each
Description
The Functional Beads can be used in conjunction with the BD CBA Functional Bead Conjugation Buffer Set (Mfr. No. 558556) to covalently link water-soluble proteins to the surface of the beads via a sulfo-SMCC chemistry. The beads, once linked to a protein, can be used as a capture matrix for analytes of interest.
Specifications
Specifications
| Certifications/Compliance | RUO |
| Content And Storage | 4°C |
| Contents | Aqueous buffered solution containing ≤0.09% sodium azide |
| Description | Functional Bead (A6) |
| For Use With (Application) | Flow Cytometry |
| Material | Polystyrene |
| Product Type | Functional Bead (A6) |
| For Use With (Equipment) | Flow cytometer |
| No. of Tests | 6000 |
| Volume (Metric) | 0.9 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction