missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human CD121a (IL-1 RI); Flex Set, Bead B6

Product Code. 15850069
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15850069

Brand: BD 560276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Human CD121a (IL-1 RI), Standard; PE Detection Reagent; Capture Bead B6.

For measuring soluble human soluble sCD121a in serum, plasma, and cell culture supernatant samples

  • Bead-based immunoassay
  • Also known as IL-1 Receptor type-1
  • Human reactivity determined by testing samples with BD CBA Human Soluble CD121a (IL-1 RI) Flex Set

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD121alpha (IL-1 RI)
Cross Reactivity Human
Includes Human CD121a (IL-1 RI), standard, PE detection reagent, capture bead B6
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human soluble CD121a (IL-1 RI) flex set (bead B6)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.