missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human IL-5 Enhanced Sensitivity Flex Set (Bead A6)

Product Code. 15851089 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15851089 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Product Code. 15851089 Supplier BD Supplier No. 561511
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Includes

  • Kit contents:
  • Enhanced Sensitivity Capture Bead A6: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
  • Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
  • Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150

Measures human Interleukin-5 (IL-5) in serum, plasma and cell culture supernatant samples

This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.

  • Bead-based immunoassay
  • Reactivity: human
  • Standard assay range: 274 to 200000fg/mL

Flow cytometry/Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Lyophilized in an aqueous buffered solution containing BSA™ and ProClin™ 150. Store at 4°C.
Includes Capture beads, detection reagent, standards
For Use With (Application) Used in conjunction with BD™ cytometric bead array (CBA)
No. of Tests 100
Quantity 100 Tests
For Use With (Equipment) Flow Cytometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.