missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-10; Flex Set, Bead C4

Product Code. 15869288
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15869288

Brand: BD 558300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Mouse IL-10; Standard; Mouse IL-10; PE Detection Reagent; Mouse IL-10; Capture Bead C4

For detecting mouse interleukins in serum and cell culture samples

The BD CBA Mouse IL-10 Flex Set is a bead based immunoassay capable of measuring mouse interleukin-10 (IL-10) in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse IL-10 Flex Set.

  • Reactivity: Mouse
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array Software

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Cross Reactivity Mouse
Includes Mouse IL-10, standard, Mouse IL-10, PE detection reagent, Mouse IL-10, capture bead C4
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse IL-10 flex set (bead C4)
compliance-icons
Product Identifier
  • Mouse IL-10 Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.