missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse IL-6; Flex Set, Bead B4
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Mouse IL-6; Standard; Mouse IL-6; PE Detection Reagent; Mouse IL-6; Capture Bead B4
The BD CBA Mouse IL-6 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-6 (IL-6) in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse IL-6 Flex Set.
- Reactivity: Mouse
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Specifications
Specifications
| Antigen | IL-6 |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-6, standard, Mouse IL-6, PE detection reagent, Mouse IL-6, capture bead B4 |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse IL-6 flex set (bead B4) |
Safety and Handling
Product Identifier
- Mouse IL-6 Flex Set
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : CMIT/MIT
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction