missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse MIP-1α; Flex Set, Bead C7
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Mouse MIP-1α, Standard (two vials), Mouse MIP-1α PE Detection Reagent (one vial), Mouse MIP-1α Capture Bead C7 (one vial)
- Mouse reactivity was determined by testing samples with Flex Set
- Theoretical limit of detection is 2.3 pg/mL
- Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow cytometry
Specifications
Specifications
| Antigen | MIP-1alpha |
| Cross Reactivity | Mouse |
| Includes | Mouse MIP-1a, standard (2 vials), Mouse MIP-1a PE detection reagent (one vial), Mouse MIP-1a capture bead C7 (one vial) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse MIP-1alpha flex set (bead C7) |
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction