missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse Th1/Th2/Th17 Cytokine standards
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
Description
- Contains a mixture of individually lyophilized recombinant proteins made up of recombinant Mouse Interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, Interferon-γ (IFN-γ), and Tumor Necrosis Factor (TNF)
Flow Cytometry
Specifications
Specifications
| Cross Reactivity | Mouse |
| Includes | Mouse interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, interferon (IFN) and tumor necrosis factor (TNF) |
| Quantity | 1 Vial |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry, Immunoassay |
| Product Type | Mouse Th1/Th2/Th17 Cytokine standard |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction