missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Rat IL-10; Flex Set, Bead A8

Product Code. 15809298
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15809298

Brand: BD 558306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Rat IL-10; Standard; Rat IL-10; PE Detection Reagent; Rat IL-10; Capture Bead A8

For detection of rat interleukins in serum and cell culture samples

The BD CBA Rat IL-10 Flex Set is a bead-based immunoassay capable of measuring rat interleukin-10 (IL-10) in serum and cell culture supernatant samples. Rat reactivity was determined by testing samples with the BD CBA Rat IL-10 Flex Set.

  • Reactivity: Rat
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 10000pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Cross Reactivity Rat
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Serum, Cell Culture Sample
Content And Storage 4°C
For Use With (Application) Flow cytometry and for detection of Rat interleukins
Product Type Rat IL-10 flex set (bead A8)

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.