missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Rat IL-2 Enhanced Sensitivity Flex Set (Bead E5)
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
- Rat IL-2 Standard
- Rat IL-2 PE Protection Reagent
- Rat IL-2 Capture Bead E5
- Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
- Protein concentration of 10,000 pg/ml (when reconstituted in 0.4 ml Assay Diluent)
Flow Cytometry
Specifications
Specifications
| Antigen | IL-2 |
| Cross Reactivity | Rat |
| Includes | 1 Vial capture bead, 1 vial detection reagent and 2 vials standard |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Measures Rat interleukin-2 (IL-2) |
| Product Type | Rat IL-2 flex set (bead E5) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction