missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Bordetella spp. Antigen and Antisera

Product Code. 11712774
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11712774 1 mL 1mL
11702844 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code. 11712774 Supplier BD Supplier No. 223101
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Quick reactive Gold standard antisera-typing panels for Bordetella Pertusis antigen and Bordetella Parapertusis antisera

  • Product type:     Antigens and Antisera
  • Groupings: A through B products
Bordetella Pertussis Antigen
  • Sub type: Quality control antigens
  • Type: Antigens
  • Quantity: 5mL
  • Shelf Life: 730 days
  • Positive Control for Bordetella pertussis antiserum
Bordetella Parapertussis Antiserum
  • Subtype: Bordetella Antisera
  • Type: Antisera
  • Lyophilized, rehydrate to 1mL
  • Shelf Life: 965 days
  • Rabbit serum for slide agglutination

Certifications

CE marked

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Type Bordetella parapertussis Antiserum
Quantity 1 mL
compliance-icons
Product Identifier
  • Vial B. Parapertussis Antiserum
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.
  • P330-Rinse mouth.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to an appropriate treatment and disposal facility in accordancewith applicable laws and regulations, a.
Supplemental information
  • MIXTURE LIST-Contain: sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.