missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Dey/Engley (D/E) Neutralizing Broth
Description
- Dehydrated
- Culture Media
- Dey/Engley (D/E) Neutralizing Broth
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, USDA |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | D/E Neutralizing Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 10g/L, Sodium Thioglycollate 1g/L, Sodium Thiosulfate 6g/L, Sodium Bisulfite 2.5g/L, Polysorbate 80 5g/L, Lecithin 7g/L, Bromcresol Purple 0.02g/L |
| For Use With (Application) | Testing of disinfectants and sanitation efficiency |
| Packaging Type | Bottle |
| Quantity | 500 g |
Safety and Handling
Product Identifier
- D/E Neutralizing Broth
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: sodium bisulfite
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?