missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Folic Acid Assay Medium
Description
- Formulation: Vitamin assay Casamino Acids 12g/L, Dextrose 40g/L, Sodium Citrate 20g/L, L-Cystine 0.2g/L, DL-Tryptophan 0.2g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Thiamine Hydrochloride 2mg/L, Pyridoxine Hydrochloride 4mg/L, Riboflavin, Niacin 2mg, Calcium Pantothenate 400μg, p-Aminobenzoic Acid 200μg, Biotin 0.8μg, Dipotassium Phosphate 1g, Monopotassium Phosphate 1g, Magnesium Sulfate 0.4g, Sodium Chloride 20mg, Ferrous Sulfate 20mg, Manganese Sulfate 20mg
Certifications
ISO, 9000
Specifications
Specifications
| Certifications/Compliance | ISO, 9000 |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Folic Acid Assay Medium |
| Product Type | Microbiology Media |
| Formulation | Vitamin assay Casamino Acids 12g/L, Dextrose 40g/L, Sodium Citrate 20g/L, L-Cystine 0.2g/L, DL-Tryptophan 0.2g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Thiamine Hydrochloride 2mg/L, Pyridoxine Hydrochloride 4mg/L, Riboflavin 2mg/L, Niacin 2mg/L, Calcium Pantothenate 400μg/L, p-Aminobenzoic Acid 200μg/L, Biotin 0.8μg/L, Dipotassium Phosphate 1g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese Sulfate 20mg/L |
| For Use With (Application) | For determining folic acid concentration by the microbiological assay technique |
| Packaging Type | Bottle |
| Quantity | 100 g |
Safety and Handling
Product Identifier
- Folic Acid Assay Medium
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P362-Take off contaminated clothing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?