missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Lactobacilli MRS Agar

Product Code. 11723553
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11723553 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11723553 Supplier BD Supplier No. 288210

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For cultivation of lactobacilli. BD Difco™ Dehydrated Culture Media: Lactobacilli MRS Agar is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Lactobacilli MRS Agar
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF, ISO, SMD
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type MRS Agar
Product Type Agar
Formulation Proteose Peptone No. 3 10g/L, Beef extract 10g/L, Yeast extract 5g/L, Dextrose 20g/L, Polysorbate 80 1g/L, Ammonium Citrate 2g/L, Sodium Acetate 5g/L, Magnesium Sulfate 0.1g/L, Manganese Sulfate 0.05g/L, Dipotassium Phosphate 2g/L, Agar 15g/L
For Use With (Application) Cultivation of lactobacilli
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.