missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Lactobacilli MRS Broth

Product Code. 11713553
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
Unit Size:
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11713553

Brand: BD 288130

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Recommended for use in the isolation, enumeration and cultivation of Lactobacillus species. BD™ Difco™ Lactobacilli MRS Broth supports luxuriant growth of all lactobacilli from oral, fecal, dairy and other sources. Supplied in a convenient, easy-to-use dehydrated powder.

Difco™ Lactobacilli MRS Broth powder contains peptone and dextrose. These ingredients supply nitrogen, carbon and other elements necessary for growth. Polysorbate 80, acetate, magnesium and manganese provide growth factors for culturing a variety of lactobacilli. The above ingredients may inhibit the growth of some organisms other than lactobacilli.

  • Based on the formulations of deMan, Rogosa and Sharpe
  • Samples can be inoculated directly into Lactobacilli MRS Broth
  • Produces a good cultural response with: Lactobacillus delbrueckii (subsp. lactis) 7830, Lactobacillus fermentum 9338 and Lactobacillus johnsonii 11506
  • pH (5.5% solution, 25°C): 6.5 ± 0.2
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF, SMD
Color Tan
Content And Storage 2°C to 8°C
Form Dehydrated Powder
Media Type MRS Broth
Product Type Microbiology Media
Formulation Proteose Peptone No. 3 10g/L, Beef extract 10g/L, Yeast extract 5g/L, Dextrose 20g/L, Polysorbate 80 1g/L, Ammonium Citrate 2g/L, Sodium Acetate 5g/L, Magnesium Sulfate 0.1g/L, Manganese Sulfate 0.05g/L, Dipotassium Phosphate 2g/L
For Use With (Application) Cultivation of lactobacilli
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.