missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: LB Agar, Lennox
Click to view available options
:
LB Agar, Lennox; 500g
Description
- Contains 5g/L NaCl
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | LB Agar |
| Product Type | Agar |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L, Agar 15g/L |
| For Use With (Application) | Propagation and maintenance of Escherichia coli for molecular biology |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction