missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani)

Product Code. 11768902
Change view
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11768902 2000 g Pail 2kg
11778902 500 g Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11768902 Supplier BD Supplier No. 244610

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Propagate and maintain E. coli for molecular biology. BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

 

  • Contains 10g/L NaCl
  • For propagation of E. coli
  • Wide range of molecular biology applications

 

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM
Content And Storage Refrigerate
Form Dehydrated
Media Type LB Broth, Miller
Product Type Microbiology Media
Formulation Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application) Propagation and maintenance of Escherichia coli for molecular biology
Packaging Type Pail
Quantity 2000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.