missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: M 17 Broth

Product Code. 11708293
Click to view available options
:
M17 Broth; 500g
This item is not returnable. View return policy

Product Code. 11708293

Brand: BD 218561

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Selective isolation and enumeration of streptococci. BD Difco™ Dehydrated Culture Media: M 17 Broth is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • M 17 Broth

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

MPF, ISO

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF, ISO
Content And Storage Refrigerate
Form Dehydrated
Media Type M17 Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 5g/L, Soy Peptone 5g/L, Beef extract 5g/L, Yeast extract 2.5g/L, Ascorbic Acid 0.5g/L, Magnesium Sulfate 0.25g/L, Disodium-beta-glycerophosphate 19g/L
For Use With (Application) Isolating lactic streptococci from yogurt, cheese starters and other dairy product
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.