missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: M9 Minimal Salts, 5x

Product Code. 11748952
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11748952 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11748952 Supplier BD Supplier No. 248510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cultivation of recombinant strains of E. coli. BD Difco™ Dehydrated Culture Media: M9 Minimal Salts, 5x is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • M9 Minimal Salts
  • 5x 

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
Form Dehydrated
Product Type Prepared Microbiology Media
Formulation Disodium Phosphate (anhydrous) 33.9g/L, Monopotassium Phosphate 15g/L, Sodium Chloride 2.5g/L, Ammonium Chloride 5g/L
For Use With (Application) Cultivation of recombinant strains of Escherichia coli
Packaging Type Bottle
Quantity 500 g
compliance-icons
Product Identifier
  • M9 Minimal Salts
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ammonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.