missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: OF Basal Medium

Product Code. 11713453
Click to view available options
:
OF Basal Medium (Oxidation/Fermentation); 500g
This item is not returnable. View return policy

Product Code. 11713453

Brand: BD 268820

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxidation/Fermentation medium for differentiation and classification of gram-negative microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM
Content And Storage Refrigerate
Form Dehydrated
Media Type OF Basal Medium
Product Type OF Basal Medium
Formulation Pancreatic digest of Casein 2g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 0.3g/L, Bromthymol blue 0.08g/L, Agar 2g/L
For Use With (Application) Determination of oxidative and fermentative metabolism of Carbohydrate by gram negative rods on the basis of Acid reaction in either the open or closed system
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.