missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Potato Dextrose Broth

Product Code. 11738982
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11738982 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11738982 Supplier BD Supplier No. 254920
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

For cultivation of Yeasts and mold. BD Difco™ Dehydrated Culture Media: Potato Dextrose Broth is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Potato Dextrose
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type Potato Dextrose Broth
Product Type Microbiology Media
Formulation Potato Starch (from infusion) 4g/L, Dextrose 20g/L
For Use With (Application) Cultivation of Yeasts and mold
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.