missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Xylose Lysine Deoxycholate (XLD) Agar

Product Code. p-4902019
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11703513

Brand: BD 278850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Moderately selective medium recommended for isolation and differentiation of enteric pathogens, especially Shigella species

  • Formulation: Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Agar 13.5g/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, JP, MCM9, SMD, SMWW, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, JP, MCM9, SMD, SMWW, USP
Content And Storage <30°C
Form Dehydrated
Media Type XLD Agar
Product Type Agar
Formulation Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Agar 13.5g/L
For Use With (Application) Isolation and differentiation of enteric pathogens
Packaging Type Bottle
Quantity 500 g
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P321-Specific treatment (see on this label).
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to an appropriate treatment and disposal facility in accordancewith applicable laws and regulations, a.
Supplemental information
  • MIXTURE LIST-Contains : Bile, extract, Ammonium iron(III) citrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.