missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Fluorescent Antibody Reagents

Product Code. 11750234 Shop All Brady Products
Change view
Click to view available options
Product Type:
FA Bordetella Parapertussis
FA Bordetella Pertussis
Unit Size:
5mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11750234 FA Bordetella Pertussis Each
11722784 FA Bordetella Pertussis 5mL
11732784 FA Bordetella Parapertussis 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11750234 Supplier BD Supplier No. 223591
  • ONLINE EXCLUSIVE PRICE
    40.65 €
    18.24 € / Pack of 1
    Save 22.41 €
    55% Off
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light

  • Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
  • For use in direct fluorescent identification

Order Info

In 5mL vials.

Specifications

Product Type FA Bordetella Pertussis
Quantity 5 mL
For Use With (Application) Direct fluorescent antibody technique for the identification of Bordetella pertussis and Bordetella parapertussis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.