missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Neutralizing Buffer for Dehydrated Culture Media

Product Code. 11728822
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11728822 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11728822 Supplier BD Supplier No. 236210
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications:

Inactivates chlorine and quaternary ammonium compounds

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM, COMPF, SMD
Content And Storage 15°C to 30°C
Additive Neutralizing Buffer
Form Dehydrated
For Use With (Application) Dehydrated media
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.