missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Salmonella VI Antigen

Product Code. 11738323
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11738323 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
The product indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states. It also contains sodium azide which is toxic by inhalation, by skin contact, and if swallowed. After contact with skin, wash immediately with plenty of water. On disposal, flush with a large volume of water to prevent azide build-up.Packaging contains dry natural rubber
Product Code. 11738323 Supplier BD Supplier No. 221421
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The product indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states. It also contains sodium azide which is toxic by inhalation, by skin contact, and if swallowed. After contact with skin, wash immediately with plenty of water. On disposal, flush with a large volume of water to prevent azide build-up.Packaging contains dry natural rubber

Quality control testing of Difco™ Salmonella O Antisera by slide agglutination tests

  • Chemically stabilized and inactivated suspensions of known strains
  • Homologous identifying antigen: Vi
  • Each vial is sufficient for 20 slide tests
  • Sodium azide (0.1%) used as preservative

Used in quality control of Salmonella O Antisera Poly A-I and Vi, Vi

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antigen
For Use With (Application) Used in the quality control of Salmonella O Antisera Poly A-I & Vi (226641) and Vi (228271)
Quantity 1 mL
compliance-icons
Product Identifier
  • BD Difco Salmonella QC Antigen Vi
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.