missing translation for 'onlineSavingsMsg'
Learn More

BD BD FACS™ Lysing Solution 10X Concentrate

Product Code. 10141013
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10141013 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10141013 Supplier BD Supplier No. 349202
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

May be used in both lyse/wash and lyse/no-wash procedures. BD™ FACS™ Lysing Solution 10X Concentrate is appropriate for use with reagents such as BD Tritest™ or BD Simultest™ reagents and a suitably equipped flow cytometer.

BD™ FACS™ lysing solution is intended for lysing red blood cells following direct immunofluorescence staining of human peripheral blood cells with monoclonal antibodies prior to flow cytometric analysis. Whole blood cell lysis helps eliminate interfering cells to permit effective detection of lymphocytes.

  • Sufficient for 2,000 tests when used in lyse/no-wash procedures (for example, BD Tritest™)
  • Sufficient for 500 tests when used in lyse/wash procedures (for example, BD Simultest™)
  • To prepare, dilute the 10X concentrate 1:10 with room temperature (20–25°C), deionized water
  • Flash point: >64°C
  • Soluble in Water
  • For In Vitro Diagnostic Use
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Content And Storage Store in cool, dry conditions in well-sealed container.
Concentration 10X
For Use With (Equipment) Flow cytometer systems, such as FACSCanto™ and FACSCalibur™ systems
Type Lysing Solution
Includes Approximate formulation: 10% formaldehyde, 30% diethylene glycol, 3.5% methanol, 56.5% water
Quantity 100 mL
Form Liquid
compliance-icons compliance-icons
Product Identifier
  • BD FACS Lysing Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H371-May cause damage to organs.
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: diethylene glycol, formaldehyde, methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.