missing translation for 'onlineSavingsMsg'
Learn More

BD FUT-175 (Futhan)

Product Code. 15836758 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15836758 5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15836758 Supplier BD Supplier No. 552035
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic, broad-specificity protease inhibitor and an inhibitor of the classical and alternate pathways of complement activation

  • Alias: Futhan
  • Chemical name: 6-amidino-2-naphthyl p-guanidinobenzoate dimethanesulfonate
  • Molecular weight: 539.58 Da
  • Storage buffer: Lyophilized powder

Serum Stab

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Serum Stab
Content And Storage 4°C
Quantity 5 mg
Molecular Weight (g/mol) 539.58
Inhibitors FUT-175 (Futhan)
Form Lyophilized Powder

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.