missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems Incubation Containers

Product Code. 11737194
Change view
Click to view available options
Holds:
Petri Dishes (15 to 18)
Petri Dishes (30 to 33)
Size:
Large
Standard
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Size Unit Size
11737194 Petri Dishes (30 to 33) Large Each
10118924 Petri Dishes (15 to 18) Standard Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11737194 Supplier BD Supplier No. 260672
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.

Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.

  • Non-breakable, chemical resistant containers
  • Removable Petri dish racks are available (sold separately)
TRUSTED_SUSTAINABILITY

Specifications

Type EZ Incubation Container
Closure Type Easy close latches
Content And Storage 2°C to 25°C
Disposable Reusable
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Holds Petri Dishes (30 to 33)
Product Line GasPak
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.