missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets

Product Code. 10612796
Change view
Click to view available options
Type:
Sachet
Sachet w/ Indicator
For Use With (Equipment):
EZ Anaerobe Container System
EZ CO2 Container System
EZ Campy Container System
Unit Size:
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) unitSize
10612796 Sachet w/ Indicator EZ Anaerobe Container System Pack of 20
11747194 Sachet EZ Anaerobe Container System Pack of 20
11767194 Sachet EZ Campy Container System Pack of 20
11757194 Sachet EZ CO2 Container System Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10612796 Supplier BD Supplier No. 260001
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets provide catalyst-free convenience.

  • For use in both the GasPak™ EZ Containers and the GasPak™ 100 and GasPak™ 150 jars
  • Sachets require no water or catalyst for activation and are engineered to provide rapid gas generation for optimal bacterial growth
TRUSTED_SUSTAINABILITY

Specifications

Type Sachet w/ Indicator
Content And Storage 2°C to 25°C
Format Sachet
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
For Use With (Equipment) EZ Anaerobe Container System
Product Line GasPak
Quantity 20 Sachets/Box

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.