missing translation for 'onlineSavingsMsg'
Learn More

BD BV480 Mouse Anti-Human IFN- γ

Product Code. p-6904082
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15814819

Brand: BD 566176

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

IFN-γ is a potent multifunctional cytokine that is produced by several activated cell types. BD Horizon™ BV480 Mouse Anti-Human IFN- γ can exert strong regulatory influences on the proliferation, differentiation, and effector responses of B cell and T cell subsets.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Mouse Anti-Human IFN- γ
Quantity 25 Tests
For Use With (Application) Intracellular Staining (Flow Cytometry)
Cross Reactivity QC Testing: Human
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.