missing translation for 'onlineSavingsMsg'
Learn More

BD Human C3a ELISA Kit

Product Code. 15876288
Change view
Click to view available options
Quantity:
1 Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15876288 1 Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15876288 Supplier BD Supplier No. 550499
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For in vitro quantitative determination Human C3a-desArg in human EDTA plasma, serum and other biological samples

The OptEIA ELISA test is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay)

  • Utilizes monoclonal antibody specific for human C3a-desArg coated on a 96-well plate

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen C3a
Cross Reactivity Human
Includes Wash concentrate (20X), stop solution, standard/sample diluent, Enzyme concentrate Streptavidin HRP, Human C3a Antibody coated plate, detection Antibody Biotin Anti-Human C3a, Human C3a lyophilized standard, ELISA diluent, TMB one step Substrate reagent
Quantity 1 Plate
Sample Type Human EDTA Plasma, Serum, Biological Sample
Content And Storage 2°C to 8°C
For Use With (Application) In-vitro quantitative determination Human C3a-desArg, ELISA
Product Type Human C3a ELISA kit
compliance-icons compliance-icons
Product Identifier
  • Hu C3a OptEIA ELISA Kit 1Plt
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.