missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human IL-10 ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
The OptEIA™ set for human interleukin-10 (IL-10) contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human IL-10 in serum, plasma, and cell culture supernatants.
- Capture antibody: anti-human IL-10
- Detection antibody: biotinylated anti-human IL-10
- Enzyme reagent: streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standard: lyophilized recombinant human IL-10
- Standard range: 7.8 - 500 pg/mL
ELISA
Specifications
Specifications
| Concentration | 7.8 to 500pg/mL |
| Product Type | Human IL-10 ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human IL-10 in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Human IL-10 (capture) Antibody, Biotinylated anti-Human IL-10 (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-10 (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction