missing translation for 'onlineSavingsMsg'
Learn More

Human Immunoglobulin Master Buffer Kit, BD

Product Code. 15829578
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829578 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15829578 Supplier BD Biosciences Supplier No. 558683
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

  • Instrument Setup Bead F1
  • Instrument Setup Bead F9
  • Instrument Setup Bead A1
  • Instrument Setup Bead A9
  • Assay Diluent
  • Detection Reagent Diluent
  • F Wash Buffer (1X)
  • PE Instrument Setup Bead F1
  • PE Positive Control Detector
  • Capture Bead Diluent

Employs particles with discrete fluorescence intensities to detect soluble immunoglobulin

Contains all of the supporting reagents necessary to perform an assay using CBA Human Immunoglobulin Flex Set.

  • Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures
  • Buffers and instrument setup reagents provided, have been optimized for analysis of immunoglobulin in serum samples.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
No. of Tests 100.0
Includes 2 bottles 40mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 260mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9, 1 vial, 1.0 mL instrument setup bead F1
Quantity 100 Tests
Validated Application RUO
Product Type Cytometric Bead Array (CBA) Assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.