Learn More
BD Human Regulatory T Cell Sorting Kit
Product Code. 15890459
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. Quantity Unit Size
15890459 1 Kit Each
1 options
Product Code. 15890459 Supplier BD Supplier No. 560753
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Human Regulatory T Cell Sorting Cocktail (CD4 PerCP-Cy5.5, CD25 PE, CD127 Alexa Fluor™ 647), CD45RA FITC, Compensation Controls
Provides reagents that allow investigators to stain viable human peripheral blood mononuclear cells (PBMC)
Flow Cytometry, Fluorescence Activated Cell Sorting
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Research Use Only