missing translation for 'onlineSavingsMsg'
Learn More

BD MicroFlow™ Mouse Micronuclei Plus Kit

Product Code. 10158335 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
60 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10158335 60 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10158335 Supplier BD Supplier No. 552730
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Anticoagulant/diluent, buffer solution, long-term storage solution (LTSS), RNase solution, mouse anti-CD71 antibody, DNA stain, platelet antibody, biological standards (positive control samples, negative control samples, CD71-negative setup samples and Malaria Biostandard blood samples), CD-ROM

For preparing mouse blood samples for flow cytometric enumeration of micronucleated erythrocyte populations

  • Reactivity: mouse
  • Size: 60 tests

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RUO
Content And Storage -75° to -85°C
Includes Anticoagulant/diluent, buffer solution, long-term storage solution (LTSS), RNase solution, mouse Anti-CD71 antibody, DNA stain, platelet antibody, biological standards, CD-ROM
For Use With (Application) For preparing mouse blood samples
No. of Tests 60
Quantity 60 Tests
For Use With (Equipment) Flow Cytometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.