Learn More
BD MicroFlow™ Rat Micronuclei Analysis Plus Kit
Product Code. 15816848
Change view
Click to view available options
Quantity:
60 Tests
Unit Size:
1 set
Product Code. Quantity Unit Size
15816848 60 Tests 1 set
1 options
Product Code. 15816848 Supplier BD Supplier No. 552731
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used when preparing rat blood samples for flow cytometric enumeration of micronucleated erythrocyte populations
MicroFlow method components are the biological standards which aid flow cytometer configuration for the micronucleus scoring application.
  • Offers advantages over other automated methods of analysis
  • Has significant advantages compared to tradition al microscopic scoring techniques
TRUSTED_SUSTAINABILITY
Specifications
Host Species Rat
Quantity 60 Tests