missing translation for 'onlineSavingsMsg'
Learn More

Mouse Foxp3 buffer set, BD

Product Code. 15820179
Change view
Click to view available options
Quantity:
Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15820179 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Irritating to eyes and skin. Do not breathe vapor. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing
Product Code. 15820179 Supplier BD Biosciences Supplier No. 560409

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Irritating to eyes and skin. Do not breathe vapor. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing

Includes

Mouse Foxp3 fixation concentrate (20 x), mouse Foxp3 permeabilization concentrate (5 x)

For intracellular staining of mouse Foxp3 and surface staining of appropriate CD markers

  • Optimized for use with Foxp3 mAb clone MF23
  • Acquire at least 15,000 to 25,000 CD4 positive lymphocytes

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mouse Foxp3 Buffer set
For Use With (Application) Intracellular staining of Mouse Foxp3, surface staining of appropriate CD markers, for Foxp3 mAb clone MF23
Quantity Ea.
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mouse Foxp3 Buffer Set
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : formaldehyde

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.