missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse T Lymphocyte Subset Antibody Cocktail

Designed to identify major subsets of T lymphocytes by direct immunofluorescent staining with flow cytometric analysis

Brand:  BD 558391

 View more versions of this product

Product Code. 10779856

  • ONLINE EXCLUSIVE PRICE
    1020.00 €
    951.23 € / 1 set
    Save 68.77 €
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The three antibodies have been titrated and pre-diluted, mixed together, and formulated for optimal staining performance. The Mouse T Lymphocyte Subset Isotype Control contains equivalent concentrations of fluorochrome- and isotype-matched negative-control immunoglobulin

  • Permits recognition of each of three antigens on each cell in a sample
  • Levels of expression of three antigens distinguish the major subpopulations of developing and peripheral T lymphocytes
  • Additional fluorochrome-labeled reagents may be combined to further characterize T-cell subpopulations
  • Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography
  • Clone: 145-2C11, RM4-5, 53-6.7
  • Isotype: Armenian Hamster IgG1, κ, Rat IgG2a, κ

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
FITC/PE/PE-Cyanine7
Affinity Purified
RUO
Cocktail
Mouse
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.