missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse T Lymphocyte Subset Antibody Cocktail

Product Code. 15849378 Shop All BD Biosciences Products
Change view
Click to view available options
Conjugate:
APC/PE/PE-Cyanine7
FITC/PE/PE-Cyanine7
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conjugate Unit Size
15849378 APC/PE/PE-Cyanine7 Each
10779856 FITC/PE/PE-Cyanine7 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15849378 Supplier BD Supplier No. 558431
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to identify major subsets of T lymphocytes by direct immunofluorescent staining with flow cytometric analysis

The three antibodies have been titrated and pre-diluted, mixed together, and formulated for optimal staining performance. The Mouse T Lymphocyte Subset Isotype Control contains equivalent concentrations of fluorochrome- and isotype-matched negative-control immunoglobulin

  • Permits recognition of each of three antigens on each cell in a sample
  • Levels of expression of three antigens distinguish the major subpopulations of developing and peripheral T lymphocytes
  • Additional fluorochrome-labeled reagents may be combined to further characterize T-cell subpopulations
  • Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography
  • Clone: 145-2C11, RM4-5, 53-6.7
  • Isotype: Armenian Hamster IgG1, κ, Rat IgG2a, κ

TRUSTED_SUSTAINABILITY

Specifications

Classification Monoclonal
Conjugate APC/PE/PE-Cyanine7
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.