missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse TCR V β Screening Panel

Product Code. 15898808
Change view
Click to view available options
Conjugate:
FITC
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conjugate Unit Size
15898808 FITC 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15898808 Supplier BD Supplier No. 557004
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monoclonal antibodies for T-cell receptors recognition

Recognizes mouse VΒ 2, 3, 4, 5.1 and 5.2, 6, 7, 8.1 and 8.2, 8.3, 9, 10b, 11, 12, 13, 14, and 17a T-cell receptors

  • Consists of 15 pre-diluted FITC-conjugated monoclonal antibodies
  • One 1mL vial of each antibody, sufficient reagents for 50 tests
  • Used at 20 microliters per 106 cells for immunofluorescent staining
  • Recommended multicolor staining to distinguish T lymphocytes from non-T cells

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC
Host Species Mouse

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.