Learn More
BD Mouse Th17/Treg Phenotyping Kit
Product Code. 15810479
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. Quantity Unit Size
15810479 1 Kit Each
1 options
Product Code. 15810479 Supplier BD Supplier No. 560767
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
CD4 PerCP-Cy5.5, IL-17A PE, Foxp3 Alexa Fluor 647 antibody cocktail, Foxp3 Buffer Set, BD GolgiStop Protein Transport Inhibitor
Simultaneously view TH17 and regulatory T cells in mouse model systems
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For research use only. Not for diagnostic or therapeutic procedures.