missing translation for 'onlineSavingsMsg'
Learn More

BD PBX Calcium Assay Kit

Product Code. 15896279
Change view
Click to view available options
Quantity:
10 Preps
100 Preps
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896279 10 Preps 1 set
15816289 100 Preps 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15896279 Supplier BD Supplier No. 640175
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Calcium Indicator, 10X PBX Signal Enhancer

For homogeneous measurement of intracellular calcium changes

  • Involves one dye addition step
  • Does not require washing
  • Allows easy implementation in highthroughput environment
  • Probenecid, a standard inhibitor of nonspecific anion transport, is not required for dye-loading
  • Can improve retention of the dye in cells

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
For Use With (Application) For homogeneous measurement of intracellular calcium changes
Includes 1 vial calcium indicator,10mL PBX signal enhancer (10X, lyophilized), 90mL calcium assay buffer 1X
Quantity 10 Preps
Type Calcium Assay Kit

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.