missing translation for 'onlineSavingsMsg'
Learn More

BD Violet Live Cell Caspase Probe

Product Code. 15874369 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874369 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15874369 Supplier BD Supplier No. 565521
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cells with active caspases can be detected. BD Pharmingen™ Violet Live Cell Caspase Probe contains a fluorochrome-labeled caspase inhibitor. Fluorochrome-labeled caspase inhibitors have three functional domains: fluorochrome , 3-amino acid sequence (VAD) that binds to the active site of the activated caspase, and fluoromethyl ketone (FMK) moiety that allows the probe to irreversibly bind the active caspase.

  • Violet Live Cell Caspase Probe is excited by the violet laser (with an excitation maximum of 403nm), and has a fluorescence emission maximum of 454nm.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 Tests
Reactivity Human, Mouse
Product Type Violet Live Cell Caspase Probe
Content And Storage -20°C
For Use With (Application) Flow Cytometry, Intracellular Staining(Flow Cytometry)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.