missing translation for 'onlineSavingsMsg'
Learn More

BD Pharmingen™ Streptavidin-HRP

Product Code. 15846438
Click to view available options
Quantity:
50 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15846438

Brand: BD 550946

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prediluted for convenient use

  • Tested by immunohistochemistry
  • Storage Buffer: Aqueous buffered solution containing BSA and ProClin™ 150 as preservative

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Conjugate HRP
Quantity 50 mL
Content And Storage 4°C
Form Liquid
For Use With (Application) Immunohistochemistry
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.