Learn More
BD Phosflow™ Human Monocyte/NK Cell Activation Kit
Product Code. 15821639
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
Product Code. Quantity Unit Size
15821639 50 Tests Each
1 options
Product Code. 15821639 Supplier BD Supplier No. 562089
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Antibody cocktail, fluorochrome-labled antibodies to six key phosphoproteins, optimized protocols and buffers
Study monocyte and NK cell signaling events without prior isolation of cells from whole blood
Complete system for the determination of the activation status of six different key phosphoproteins in B cells, monocytes, NK cells, and T cells using whole blood samples.
Evaluate:
  • Phosphorylation status information on the intracellular MAPK and JAK/STAT signaling pathway
  • Responses transduced by treated human monocytes and NK cells
  • Signaling events in T and B-cell populations
Benefits:
  • Lyophilized positive and negative control cells to increase confidence and standardization
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Host Species Human
Quantity 50 Tests
Product Type Human Monocyte/NK Cell Activation Kit