missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosflow Lyse/Fix Buffer 5X, BD
Descripción
- Can be used for lysing red blood cells and for fixing leukocytes in one step, allowing investigators to rapidly inactivate kinases, phosphatases and/or proteases, so that the in vivo phosphorylation state of the cell can be examined
- Has been reported to preserve the light scattering properties of the cell
- May be used on human and mouse whole blood
- Whole blood lysis has been shown to be as effective as density gradient centrifugation in the preparation of PBMCs for lymphocyte subset analysis
- In many cases, phospho-specific antibodies may be used simultaneously with the staining of cell surface markers (e.g. CD markers); however, not all cell surface markers, however, are compatible with the BD Phosflow Lyse/Fix Buffer
Especificaciones
Especificaciones
| Color | Clear |
| Content And Storage | Room Temperature |
| Format | Liquid |
| Quantity | 250mL |
| Validated Application | RUO |
| Product Type | Lyse/Fix Buffer |
Seguridad y manipulación
Nombre del producto
- BD Phosflow Lyse/Fix Buffer
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 2
- H241-Heating may cause a fire or explosion.
- H371-May cause damage to organs.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contain: formaldehyde, diethylene glycol, methanol
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?