missing translation for 'onlineSavingsMsg'
Learn More

Phosflow Perm Buffer II, BD

Product Code. 15869178
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15869178 125 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869178 Supplier BD Biosciences Supplier No. 558052
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For the permeabilization of fixed Peripheral Blood Mononuclear Cells (PBMCs) or lysed whole blood.

  • Designed for optimal fluorescent staining in flow cytometry
  • Can be used with antibodies for phospho-protein analysis
  • In many cases, may be used simultaneously with phospho-specific antibodies and the staining of cell surface markers (e.g CD markers)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Liquid
Quantity 125 mL
Validated Application RUO
Product Type Staining Buffer
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BD Phosflow Perm Buffer II
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.