missing translation for 'onlineSavingsMsg'
Learn More

BD Phospholipid-binding Protein, APC Annexin V

Product Code. 15866278 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
200 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15866278 200 Tests Each
15856278 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15866278 Supplier BD Supplier No. 550475
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Has high affinity for PS, and binds to cells with exposed PS

  • Conjugated to APC
  • Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 Tests
Sample Volume 5 μL
Storage Buffer Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Conjugate APC (Allophycocyanin)
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.