missing translation for 'onlineSavingsMsg'
Learn More

PI/RNase Staining Buffer, BD

Product Code. 15876398
Change view
Click to view available options
Quantity:
100mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876398 100mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15876398 Supplier BD Biosciences Supplier No. 550825
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

May be used to analyze cell cycle by flow cytometry or used with antibodies to examine the expression of proteins during the cell cycle

Propidium Iodide (PI) is a fluorescent vital dye that stains DNA and RNA. PI binds to both DNA and RNA, so the latter must be removed by digestion with ribonucleases. The content of DNA as determined by flow cytometry, can reveal useful information about the cell cycle and the proteins involved in the regulation of the cell cycle. Cells in G2 and M phases of the cell cycle have double the DNA content of those in G0 and G1 phases. Cells in S phase have a DNA content lying between these extremes.

  • PI is detected using a 562-588nm band pass filter
  • Storage Buffer: Aqueous buffered solution containing ≤0.09% sodium azide

TRUSTED_SUSTAINABILITY

Specifications

Color Pink
Content And Storage 4°C
Format Liquid
Quantity 100mL
Validated Application RUO
Product Type Staining Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.